missing translation for 'onlineSavingsMsg'
Learn More

KCNK9 Antibody, Novus Biologicals™

Product Code. 18225751 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18225751 0.025 mL 25µL
18222261 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18225751 Supplier Novus Biologicals Supplier No. NBP1464930.025ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KCNK9 Polyclonal specifically detects KCNK9 in Mouse samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK9
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500
Formulation Whole antisera with No Preservative
Gene Alias KT3.2, potassium channel, subfamily K, member 9, TASK-3
Gene Symbols KCNK9
Host Species Rabbit
Immunogen A synthetic peptide from the c-terminal region of mouse KCNK9 (TASK3) conjugated to an immunogenic carrier protein was used as the immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51305
Test Specificity Specific for KCNK9.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.