Learn More
Alfa Aesar™ Karl Fischer Coulometric AG-Oven, anolyte solution, for use with oven
Product Code. 15447445
Click to view available options
Quantity:
6 x 500 mL
Unit Size:
Each
Product Code. 15447445
Brand: Alfa Aesar™ 047177.BA
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Chemical Name or Material Karl Fischer Coulometric AG-Oven
Name Note anolyte solution, for use with oven
Boiling Point 65°C
Color Colorless to Yellow
Density 0.970g/mL
Physical Form Liquid
Quantity 6 x 500 mL
UN Number UN3286