missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 3/PSA Antibody (CL9414) [Alexa Fluor™ 700], Novus Biologicals™
Shop All Bio Techne ProductsDescription
Kallikrein 3/PSA Monoclonal antibody specifically detects Kallikrein 3/PSA in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Kallikrein 3/PSA |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL9414 |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P07288, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?