missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kaiso Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35285-20ul
This item is not returnable.
View return policy
Description
Kaiso Polyclonal antibody specifically detects Kaiso in Human,Mouse,Rat samples. It is validated for ChIP assay,ELISA,Western Blot
Specifications
| Kaiso | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, Chromatin Immunoprecipitation 5ug antibody for 10ug-15ug of Chromatin, ELISA | |
| KAISOkaiso transcription factor, transcriptional regulator Kaiso, WUGSC:H_DJ525N14.1, zinc finger and BTB domain containing 33, Zinc finger and BTB domain-containing protein 33, ZNF348kaiso, ZNF-kaiso | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 523-672 of human Kaiso (NP_006768.1).,, Sequence:, PCRYCEKVFPLAEYRTKHEIHHTGERRYQCLACGKSFINYQFMSSHIKSVHSQDPSGDSKLYRLHPCRSLQIRQYAYLSDRSSTIPAMKDDGIGYKVDTGKEPPVGTTTSTQNKPMTWEDIFIQQENDSIFKQNVTDGSTEFEFIIPESY | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ChIP Assay, ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10009 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction