missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ JTE 907

Product Code. 18034952
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18034952 10 mg 10mg
18034492 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18034952 Supplier R&D Systems™ Supplier No. 2479/10
  • ONLINE EXCLUSIVE PRICE
    314.00€
    296.10€ / 10mg
    Save 17.90€
    5% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective CB2 receptor inverse agonist

Highly selective cannabinoid CB2 receptor inverse agonist. Binds with high affinity to rat, mouse and human CB2 receptors (Ki values are 0.38, 1.55 and 35.9 nM respectively). Produces anti-inflammatory effects in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 282089-49-0
Molecular Formula C24H26N2O6

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 438.48
Purity >99%
Molecular Weight (g/mol) 438.48
Chemical Name or Material JTE 907
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.