accessibility menu, dialog, popup

UserName

H3COHNOONHOOOCH3

CAS RN 282089-49-0

IUPAC Name: N-[(2H-1,3-benzodioxol-5-yl)methyl]-7-methoxy-2-oxo-8-(pentyloxy)-1,2-dihydroquinoline-3-carboxamide
Synonyms: N-(2H-1,3-benzodioxol-5-ylmethyl)-7-methoxy-2-oxo-8-(pentyloxy)-1H-quinoline-3-carboxamide
Molecular Weight (g/mol): 438.48
Molecular Formula: C24H26N2O6
InChi Key: GRAJFFFXJYFVOC-UHFFFAOYSA-N
SMILES: CCCCCOC1=C2NC(=O)C(=CC2=CC=C1OC)C(=O)NCC1=CC2=C(OCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results