missing translation for 'onlineSavingsMsg'
Learn More

Abcam JSH 23, NFkappaB Transcriptional Activity Inhibitor, Solid, 25 mg

Product Code. 32109445
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109445 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109445 Supplier Abcam Supplier No. ab14482425mg
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent NFκB transcriptional activity inhibitor that inhibits nuclear translocation without affecting IκB degradation.

  • Molecular Weight: 240.34 Da
  • Purity: >99%
  • Form: Solid
  • Activity: NFκB transcriptional activity inhibitor (IC50 = 7.1 μM)
  • Mechanism: Inhibits LPS-induced nuclear translocation of NFκB; does not affect IκB degradation
  • Applications: Potentiates the antitumor effects of cisplatin (ab141398); shows neuroprotective effects in vivo
  • Storage: Store at -20°C
  • Shipping: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 99.97% JSH 23
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Target Molecule NFκB
Formulation Constituents: 99.97% JSH 23
Quantity 25 mg
For Use With (Application) NFκB transcriptional activity inhibition, antitumor research, neuroprotective research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.