missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD6/PSR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57068-25ul
This item is not returnable.
View return policy
Description
JMJD6/PSR Polyclonal specifically detects JMJD6/PSR in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| JMJD6/PSR | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL | |
| bifunctional arginine demethylase and lysyl-hydroxylase JMJD6, EC 1.14.11, EC 1.14.11.-, Histone arginine demethylase JMJD6, JmjC domain-containing protein 6, jumonji domain containing 6, Jumonji domain-containing protein 6, KIAA0585PSR, Lysyl-hydroxylase JMJD6, Peptide-lysine 5-dioxygenase JMJD6, Phosphatidylserine receptorPTDSR, Protein PTDSR, PTDSR1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| JMJD6 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:STPRELIKVTRDEGGNQQDEAITWFNVIYPRTQLPTWPPEFKPLEILQKPGETVFVPGGWWHVVLNLDTTIAITQNFASSTNFPVVWHKTVRGRPKLSRKWYRILKQEHPELAVLA | |
| 25 μL | |
| Epigenetics, Stem Cell Signaling Pathway | |
| 23210 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction