missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JMJD1A Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149243
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
JMJD1A is a histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a central role in histone code. It preferentially demethylates mono- and dimethylated H3 'Lys-9' residue, with a preference for dimethylated residue, while it has weak or no activity on trimethylated H3 'Lys-9'. Demethylation of Lys residue generates formaldehyde and succinate. JMJD1A is involved in hormone-dependent transcriptional activation, by participating in recruitment to androgen-receptor target genes, resulting in H3 'Lys-9' demethylation and transcriptional activation. It's also Involved in spermatogenesis by regulating expression of target genes such as PRM1 and TMP1 which are required for packaging and condensation of sperm chromatin. Involved in obesity resistance through regulation of metabolic genes such as PPARA and UCP1.
Specifications
| JMJD1A | |
| Polyclonal | |
| Unconjugated | |
| KDM3A | |
| 1700105C21Rik; C230043E16Rik; DKFZp686A24246; DKFZp686P07111; JHDM2a; JHMD2A; JmjC domain-containing histone demethylation protein 2A; Jmjd1; Jmjd1a; jumonji C domain-containing histone demethylase 2A; jumonji domain containing 1A; jumonji domain-containing protein 1A; KDM2A; Kdm3a; Kiaa0742; lysine (K)-specific demethylase 3A; lysine demethylase 3A; lysine-specific demethylase 3A; probable zinc finger protein; testis specific gene A; testis-specific gene A protein; testis-specific protein A; TGSA; Tsga; zinc finger protein TSGA | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 55818 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9Y4C1 | |
| KDM3A | |
| E. coli-derived recombinant human KDM3A, Pro844-Glu968. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction