missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JM4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35838-100ul
This item is not returnable.
View return policy
Description
JM4 Polyclonal antibody specifically detects JM4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| JM4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| FLJ57916, Jena-Muenchen 4, JM4, PRA1 domain family 2, PRA1 domain family, member 2, PRA1 family protein 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-50 of human JM4 (NP_009144.1).,, Sequence:, MSEVRLPPLRALDDFVLGSARLAAPDPCDPQRWCHRVINNLLYYQTNYLL | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 11230 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction