missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JIP1 Antibody (OTI1D9), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00€ - 550.00€
Specifications
| Antigen | JIP1 |
|---|---|
| Clone | OTI1D9 |
| Dilution | Immunohistochemistry 1:150 |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306586
|
Novus Biologicals
NBP3-44895-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306489
|
Novus Biologicals
NBP3-44895-30ul |
30 μL |
256.00€
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JIP1 Monoclonal antibody specifically detects JIP1 in Human,Mouse,Rat samples. It is validated for ImmunohistochemistrySpecifications
| JIP1 | |
| Immunohistochemistry 1:150 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C-Jun-amino-terminal kinase-interacting protein 1;IB1;JIP1;JIP-1;MAPK8IP1;PRKM8IP | |
| Full length human recombinant protein of human MAPK8IP1 (NP_005447) produced in HEK293T cell. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI1D9 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Signal Transduction | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 9479 | |
| IgG1 | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title