missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAG1 Monoclonal Antibody (OTI3A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807944
This item is not returnable.
View return policy
Description
The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.Specifications
| JAG1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P78504 | |
| JAG1 | |
| Human recombit protein fragment corresponding to amino acids 523-759 of human JAG1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A10 | |
| Unconjugated | |
| JAG1 | |
| AGS, AGS1, AHD, AWS, CD339, HJ1, JAGL1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 182 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction