Learn More
Avantor™ J.T. Baker™ BAKERBOND spe™ SDB-1 SPE columns, polymer based
Product Code. 10331281 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
50
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10331281 50 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10331281 Supplier Avantor™ Supplier No. 751902
  • ONLINE EXCLUSIVE PRICE
    285.42€
    256.59€ / Pack of 50
    Save 28.83€
    10% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Can improve the quality of sample preparation
  • Excellent structural, mechanical and chemical properties, packaging purity and load ability, as well as stability of the SDB sorbent, offer improved sample clean-up and pre-concentration.
  • This SDB polymeric sorbent is a highly efficient and selective SDB sorbent with high throughput performance.
  • It enables high recovery and reproducibility of results, even in the SPE of compounds of different polarities.
Specifications
Column Capacity (Metric) 3 mL
Bed Weight 200 mg
Quantity 50