missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ITPK1 (Human) Recombinant Protein

Human ITPK1 full-length ORF ( AAH03622, 1 a.a. - 314 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00003705-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107041

  • 528.00€ / 25µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

  • Sequence: MQTFLKGKRVGYWLSEKKIKKLNFQAFAELCRKRGMEVVQLNLSRPIEEQGPLDVIIHKLTDVILEADQNDSQSLELVHRFQEYIDAHPETIVLDPLPAIRTLLDRSKSYELIRKIEAYMEDDRICSPPFMELTSLCGDDTMRLLEKNGLTFPFICKTRVAHGTNSHEMAIVFNQEGLNAIQPPCVVQNFINHNAVLYKVFVVGESYTVVQRPSLKNFSAGTSDRESIFFNSHNVSKPESSSVLTELDKIEGVFERPSDEVIRELSRALRQALGVSLFGIDIIINNQTGQHAVIDINAFPGDCQVCFIEGWKTD
Specifications

Specifications

AAH03622
inositol 1,3,4-triphosphate 5/6 kinase
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ITPK1
GST
3705
Wheat germ expression system
25 μg
ITRPK1
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers