Learn More
Eppendorf™ IsoTherm™ System Starter Set
Product Code. p-8004110 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
0.5mL tubes
1,5/2,0mL tubes
Unit Size:
Each
Product Code. 10114721
Brand: Eppendorf™ 3880000011
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Holds up to 24 micro tubes. The IsoTherm-System is the ideal system for safely cooling your samples in 0.5mL or 1.5 mL/2.0 mL tube formats at -21°C or 0°C throughout your entire lab workflow.
  • IsoPack™ (cold pack) white for 0°C and blue for -21°C, cooled to refrigeration temperature in freezer compartment
  • Isothermal times of >6hr (0°C, white) and >3hr (-21°C, blue)
  • No more melting ice baths (danger of contamination)
  • All system components can be used separately
  • Racks are stackable, autoclavable, markable, floatable and can be centrifuged in a microtitre-plate rotor
  • Specializes in cooling samples effectively and constantly at -21°C or 0°C over many hours.
TRUSTED_SUSTAINABILITY
Specifications
Type IsoTherm-System Starter Set
For Use With (Equipment) 0.5mL tubes