missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Isopeptidase T/USP5 Antibody [PE], Novus Biologicals™

Product Code. 30037242 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30037242 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30037242 Supplier Novus Biologicals™ Supplier No. NB10077286PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Isopeptidase T/USP5 Polyclonal antibody specifically detects Isopeptidase T/USP5 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin),Western Blot (Negative)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Isopeptidase T/USP5
Applications Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative)
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias Deubiquitinating enzyme 5, EC 3.1.2.15, EC 3.4.19.12, Isopeptidase T, IsoT, ISOTubiquitin carboxyl-terminal hydrolase 5, ubiquitin isopeptidase T, ubiquitin specific peptidase 5 (isopeptidase T), ubiquitin specific protease 5 (isopeptidase T), ubiquitin thioesterase 5, Ubiquitin thiolesterase 5, ubiquitin-specific protease-5 (ubiquitin isopeptidase T), Ubiquitin-specific-processing protease 5
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human ubiquitin specific peptidase 5 (isopeptidase T) using the numbering given in entry CAA62690.1 (GeneID 8078).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8078
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.