Learn More
Isofenphos solution, Honeywell Fluka™
Product Code. 15979691 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15979691 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15979691 Supplier Honeywell Fluka Supplier No. R13612ML
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
100 μg/mL in acetonitrile, analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
CAS 75-05-8, 25311-71-1
Flash Point 6°C
Physical Form Liquid
Boiling Point 82°C
Industry Type Acaricides and Insecticides
Quantity 2 mL
Grade Analytical Standard