Learn More
Isofenphos-oxon 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30328717
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328717 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30328717 Supplier Dr. Ehrenstorfer Supplier No. DREA14423000AL100
 Restricted Item
Estimated Shipment: 07-12-2026
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Isofenphos-oxon
CAS 31120-85-1
Analyte or Component Names Isofenphos-oxon
Shipping Condition Room Temperature
Molecular Formula C15 H24 N O5 P
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C15H24NO5P/c1-6-19-22(18,16-11(2)3)21-14-10-8-7-9-13(14)15(17)20-12(4)5/h7-12H,6H2,1-5H3,(H,16,18)
SMILES CCOP(=O)(NC(C)C)Oc1ccccc1C(=O)OC(C)C
IUPAC Name propan-2-yl 2-[ethoxy-(propan-2-ylamino)phosphoryl]oxybenzoate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 329.33
Quantity 1 mL
Formula Weight 329.1392 g/mol