Learn More
ISO 10695:2000 Standard Mixture 366 10 μg/mL in Acetone, Dr. Ehrenstorfer
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Specifications
Specifications
| Chemical Name or Material | Atrazine∣Cyanazine∣Metazachlor∣Parathion-ethyl∣Parathion-methyl∣Pendimethalin∣Propazine∣Sebuthylazine∣Simazine∣Terbuthylazine∣Trifluralin∣Vinclozolin |
| CAS | 1912-24-9, 21725-46-2, 67129-08-2, 56-38-2, 298-00-0, 40487-42-1 |
| Analyte or Component Names | Atrazine∣Cyanazine∣Metazachlor∣Parathion-ethyl∣Parathion-methyl∣Pendimethalin∣Propazine∣Sebuthylazine∣Simazine∣Terbuthylazine∣Trifluralin∣Vinclozolin |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 10 mL |
| Concentration or Composition (by Analyte or Components) | Atrazine 10 μg/mL∣Cyanazine 10 μg/mL∣Metazachlor 10 μg/mL∣Parathion-ethyl 10 μg/mL∣Parathion-methyl 10 μg/mL∣Pendimethalin 10 μg/mL∣Propazine 10 μg/mL∣Sebuthylazine 10 μg/mL∣Simazine 10 μg/mL∣Terbuthylazine 10 μg/mL∣Trifluralin 10 μg/mL∣Vinclozolin 10 μg/mL |