Learn More
IRX6 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24440-1-AP-20UL
Additional Details : Weight : 0.00020kg
Description
The IRX6 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human IRX6. This antibody recognizes human, mouse antigen. The IRX6 antibody has been validated for the following applications: IHC, WB,ELISA analysis.
Specifications
IRX6 | |
Polyclonal | |
Unconjugated | |
20 μL | |
79190 | |
Liquid |
Western Blot, Immunohistochemistry, ELISA | |
800 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |