missing translation for 'onlineSavingsMsg'
Learn More

Irisin/FNDC5 Antibody, FITC, Novus Biologicals™

Product Code. 18203275 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18203275 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18203275 Supplier Novus Biologicals Supplier No. NBP259680F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Irisin/FNDC5 Polyclonal specifically detects Irisin/FNDC5 in Human, Mouse, Camel samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Irisin/FNDC5
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS with 0.05% Sodium Azide
Gene Alias fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FRCP2Fibronectin type III repeat-containing protein 2
Gene Symbols FNDC5
Host Species Rabbit
Immunogen Partial recombinant protein made to an internal region of the human Irisin/FNDC5 protein (amino acids 32-143) [UniProt Q8NAU1]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 252995
Target Species Human, Mouse, Camelid
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.