Learn More
Iridium standard solution, for AAS, 1 mg/ml Ir in 5% HCl, Thermo Scientific Chemicals
Product Code. p-4509778 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 10480471
Brand: Thermo Scientific Acros 196041000
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Iridium standard solution
Packaging Plastic bottle
CAS 13569-57-8, 7647-01-0, 7732-18-5
Density 1.0200g/mL
Specific Gravity 1.02
Physical Form Solution
Molecular Formula Cl3H6IrO3
Concentration 990 to 1010 μg/mL of Ir(3+) in 10 to 20% HCl
Color Pink
Melting Point -25.0°C
Boiling Point 107.0°C
Solubility Soluble in water
InChI Key LNJXVUXPFZKMNF-UHFFFAOYSA-K
SMILES O.O.O.Cl[Ir](Cl)Cl
IUPAC Name trichloroiridium;trihydrate
Molecular Weight (g/mol) 352.612
PubChem CID 6102153
Quantity 100 mL
Grade AAS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iridium atomic absorption standard solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.