Learn More
IREM-1 (CD300f) Mouse anti-Human, BUV496, Clone: UP-D1, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 750018
This item is not returnable.
View return policy
Description
IREM-1 (CD300f) Monoclonal antibody specifically detects IREM-1 (CD300f) in Human samples. It is validated for Flow Cytometry
Specifications
| IREM-1 (CD300f) | |
| Monoclonal | |
| Brilliant Ultraviolet 496 | |
| CD300F; CD300LF; IREM1; NKIR; CLM-1; CLM1; IGSF13 | |
| Human Recombinant IREM-1 Protein | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| UP-D1 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody. |