missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP6K1 Antibody (10611), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43667
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry (Paraffin)
IP6K1 Monoclonal specifically detects IP6K1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IP6K1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| IP6K1 | |
| Recombinant protein encompassing a sequence within the center region of human IP6K1. The exact sequence is proprietary. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 10611 | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, EC 2.7.4.21, inositol hexakisphosphate kinase 1, Inositol hexaphosphate kinase 1KIAA0263IHPK1PiUS, InsP6 kinase 1, MGC9925, Pi uptake stimulator | |
| Mouse | |
| 50 kDa | |
| 0.1 mL | |
| Neuroscience, Neurotransmission, Signal Transduction | |
| 9807 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction