Learn More
Ionisation buffer solution for AAS and ICP, Lithium 2% Li in HNO3 1%, Honeywell Fluka™
Product Code. 16664454 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16664454 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16664454 Supplier Honeywell Fluka Supplier No. E4301100ML
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ionisation buffer Solution for AAS and ICP
Density 1.01 g/cm3
pH <2
Quantity 100 mL
Grade Standards and Modifiers for AAS