missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Involucrin Antibody (SY5), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2727AF594
This item is not returnable.
View return policy
Description
Involucrin Monoclonal specifically detects Involucrin in Human, Porcine, Canine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Involucrin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| IVL | |
| Purified involucrin from human keratinocytes. | |
| 0.1 mL | |
| Primary | |
| Human, Pig, Canine, Primate | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SY5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| involucrin | |
| Mouse | |
| Protein A purified | |
| Extracellular Matrix | |
| 3713.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction