missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Assay type: Cell-based
- Detection method: Fluorescent
- Results type: Quantitative
- Sample types: Suspension cells and Adherent cells
- Recommended for use with Microplate reader
- Cited in 5 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Detection Location | In-Cell Detection |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 1000 Tests |
| Target Molecule | pH |
| Detection Method | Fluorescence |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 1000 Tests |
| For Use With (Application) | Intracellular pH assay |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?