missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin αvβ6 Mouse anti-Human, BV605, Clone: 10D5, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 751821
This item is not returnable.
View return policy
Description
Integrin αvβ6 Monoclonal antibody specifically detects Integrin αvβ6 in Human samples. It is validated for Flow Cytometry.
Specifications
| Integrin αvβ6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Hu ITGRN ALPVBTA6 BV605 10D5 50 μg | |
| 129/C57BL/6 β6 -/- Mice were immunized with Human αvβ6 or Mouse Keratinocytes | |
| RUO | |
| Integrin αvβ6 | |
| Refrigerate (2 to 8°C) | |
| Human |
| Flow Cytometry | |
| 10D5 | |
| Brilliant Violet 605 | |
| Mouse | |
| 1 x 50 μg | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction