missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin αE2 (CD103) Mouse anti-Rat, BV711, Clone: OX-62, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 745451
This item is not returnable.
View return policy
Description
Integrin αE2 (CD103) Monoclonal antibody specifically detects Integrin αE2 (CD103) in Rat samples. It is validated for Flow Cytometry.
Specifications
| Integrin αE2 (CD103) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Rat | |
| Purified |
| Flow Cytometry | |
| OX-62 | |
| Brilliant Violet 711 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction