Learn More
Integrin beta 4/CD104 Antibody (CL6972) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76524DL594
Product Code. 30298757
  • 737.00€ / 0.10mL
Description
Integrin beta 4/CD104 Monoclonal antibody specifically detects Integrin beta 4/CD104 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin beta 4/CD104
Monoclonal
DyLight 594
CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4
This antibody was developed using a recombinant protein derived from P16144, with the exact immunogen sequence remaining proprietary.
0.1 mL
Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
3691
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL6972
50mM Sodium Borate
Mouse
Protein A purified