missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 4/CD104 Antibody (450-9D) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
225.00€ - 537.00€
Specifications
| Antigen | Integrin beta 4/CD104 |
|---|---|
| Clone | 450-9D |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:500-1:1000 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18012086
|
Novus Biologicals
NB100-65599SS |
0.025 mg |
225.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18039945
|
Novus Biologicals
NB100-65599 |
0.1 mg |
537.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin beta 4/CD104 Monoclonal specifically detects Integrin beta 4/CD104 in Human, Ferret, Mustelid samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| Integrin beta 4/CD104 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| P16144 | |
| 3691 | |
| Purified alpha6 beta4 integrin from A431 cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 450-9D | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Ferret, Mustelid | |
| CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 | |
| ITGB4 | |
| IgG1 | |
| Protein G purified | |
| NB100-65599 recognizes the human beta4 integrin (also known as CD104), a 205kD glycoprotein which associates with the alpha6 integrin to form the alpha6/beta4 complex. CD104 is expressed on epithelial cells, Schwann cells and various tumour cell lines. NB100-65599 recognizes an extracellular epitope on the CD104 molecule. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts