missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 1/CD29 Antibody (K20) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52708-0.025MG
This item is not returnable.
View return policy
Description
Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| Integrin beta 1/CD29 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| ITGB1 | |
| Human T-cell lymphoma cells injected into Biozzi high-responder mice. | |
| 0.025 mg | |
| Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 3688 | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry (Frozen), CyTOF | |
| K20 | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready | |
| CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognises human CD29. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction