missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 1/CD29 Antibody (CL15606) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44157
This item is not returnable.
View return policy
Description
Integrin beta 1/CD29 Monoclonal antibody specifically detects Integrin beta 1/CD29 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
Specifications
| Integrin beta 1/CD29 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:2500-1:5000 | |
| CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
| This antibody was generated using a recombinant protein sequence of P05556, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Cancer, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 3688 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL15606 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction