missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V beta 6 Antibody (Av beta6 62ow.17), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50450F
This item is not returnable.
View return policy
Description
Integrin alpha V beta 6 Monoclonal specifically detects Integrin alpha V beta 6 in Human, Mouse (Negative) samples. It is validated for Immunohistochemistry.
Specifications
| Integrin alpha V + beta 6 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Mouse fibroblasts retrovirally infected with cDNA to human beta6 (3T3 beta6.19). Cells expressed a mouse/human av beta6 chimera. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
| Does not recognize mouse av beta 6. | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunohistochemistry | |
| Av beta6 62ow.17 | |
| Immunohistochemistry | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction