missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha E/CD103 Antibody (2G5.1), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB100-65271-0.025MG

 View more versions of this product

Product Code. 18003184

  • 204.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Integrin alpha E/CD103
Monoclonal
1.0 mg/mL
Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100
P38570
ITGAE
Human Intestinal lymphocyte
0.025 mg
Innate Immunity
3682
Human, Primate
Purified
Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Flow Cytometry
2G5.1
Unconjugated
PBS, 0.5% Bovine Serum Albumin with 0.09% Sodium Azide
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen
Mouse
Protein A purified
RUO
Primary
NB100-65271 recognizes the human CD103 cell surface antigen, also known as the alphaE integrin. This reacts with various subsets of human intra-epithelial lymphocytes:- 40% of lamina propria T-cells; 30% of mesenteric lymphoblasts and some lymphocytes from other mucosa. It recognizes less than 0.1% bone marrow cells and less than 0.7% PBL.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only