Learn More
Novus Biologicals™ Integrin alpha 5 beta 1 Antibody (BMS patent anti-Integrin alpha5beta1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28423-1mg
 View more versions of this product
Product Code. 30044133
  • 920.00€ / 1mg
Description
Integrin alpha 5 beta 1 Monoclonal antibody specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha 5 beta 1
Monoclonal
Unconjugated
CD49 antigen-like family member E, CD49e, Fibronectin receptor subunit alpha, fibronectin receptor, alpha polypeptide, fibronectin receptor, alpha subunit, FNRA, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5/SLC9B1 fusion, very late activation protein 5, alpha subunit, VLA-5, VLA5A
Integrin a5b1 (ITGA5 & ITGB1)
1 mg
Angiogenesis, Cancer, Immunology
3678
Human
Purified
Flow Cytometry, ELISA, Functional Assay
BMS patent anti-Integrin alpha5beta1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.