missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 2/CD49b Antibody (CL7318) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76483AF594
This item is not returnable.
View return policy
Description
Integrin alpha 2/CD49b Monoclonal antibody specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Integrin alpha 2/CD49b | |
| Monoclonal | |
| Alexa Fluor 594 | |
| alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2 | |
| This antibody was developed using a recombinant protein derived from P17301, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cellular Markers, Innate Immunity, Signal Transduction | |
| 3673 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL7318 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction