missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 2 beta 1 Antibody (vatelizumab), CoraFluor™ 1, Novus Biologicals™

Product Code. 30154074
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30154074 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30154074 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328512CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

Integrin alpha 2 beta 1 Monoclonal antibody specifically detects Integrin alpha 2 beta 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha 2 beta 1
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone vatelizumab
Conjugate CoraFluor 1
Formulation PBS
Host Species Human
Immunogen Integrin a2b1 (ITGA2 & ITGB1)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.