Learn More
Integrin alpha 1/CD49a Antibody (CL7217) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76479MFV610
Product Code. 30298590
  • 780.00€ / 0.10mL
Description
Integrin alpha 1/CD49a Monoclonal antibody specifically detects Integrin alpha 1/CD49a in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha 1/CD49a
Monoclonal
mFluor Violet 610 SE
CD49 antigen-like family member A, CD49a, CD49a antigen, integrin, alpha 1, Laminin and collagen receptor, very late activation protein 1, VLA-1, VLA1integrin alpha-1
This antibody was developed using a synthetic peptide derived from P56199, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
3672
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL7217
50mM Sodium Borate
Mouse
Protein A purified