missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin β7 Chain Rat anti-Mouse, BUV737, Clone: M293, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 748623
This item is not returnable.
View return policy
Description
Integrin β7 Chain Monoclonal antibody specifically detects Integrin β7 Chain in Mouse samples. It is validated for Flow Cytometry
Specifications
| Integrin β7 Chain | |
| Monoclonal | |
| Brilliant Ultraviolet 737 | |
| Itgb7; Integrin βp; Integrin beta-P; Integrin beta-7; Ly69; M290 IEL Ag | |
| αIELβ7 integrin from the C57BL/10 mouse T-cell hybridoma MTC-1 | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| M293 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction