missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (E2-E3 + 2D11-H5 (same as INS04 + INS05)) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45107
This item is not returnable.
View return policy
Description
Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse, Porcine, Bovine, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Insulin | |
| Monoclonal | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig, Bovine, Rabbit | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| E2-E3 + 2D11-H5 (same as INS04 + INS05) | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 0.1 - 0.2 μg/mL | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Purified pig insulin, conjugated to KLH (E2-E3 & 2D11-H5) (Uniprot: P01308) | |
| 7 mL | |
| Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers | |
| 3630 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction