Learn More
INSM1 Antibody (CL11630) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-15120PE
Product Code. 30300110
  • 737.00€ / 0.10mL
Description
INSM1 Monoclonal antibody specifically detects INSM1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
INSM1
Monoclonal
PE
IA1IA-1, insulinoma-associated 1, insulinoma-associated protein 1, Zinc finger protein IA-1
This antibody was developed using a synthetic peptide derived from Q01101, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
3642
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL11630
PBS
Mouse
Protein A purified