missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSIG-1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-55244H
This item is not returnable.
View return policy
Description
INSIG-1 Polyclonal specifically detects INSIG-1 in Human, Rat samples. It is validated for Western Blot.
Specifications
| INSIG-1 | |
| Polyclonal | |
| Western Blot | |
| O15503 | |
| INSIG1 | |
| A synthetic peptide representing an internal region of the human INSIG-1 protein (between residues 1-100) according to Swiss-Prot# O15503. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific for Insig1 but not Insig2. | |
| Store at 4C in the dark. |
| Western Blot | |
| HRP | |
| PBS with No Preservative | |
| CL6, CL-6, INSIG-1, INSIG-1 membrane protein, insulin induced gene 1, insulin-induced gene 1 protein, MGC1405 | |
| Rabbit | |
| 30 kDa | |
| 0.1 mL | |
| Cholesterol Metabolism, ER Markers, Lipid and Metabolism, Signal Transduction | |
| 3638 | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction