missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iNOS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00€ - 326.00€
Specifications
| Antigen | iNOS |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 1:300-1:10000 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18628945
|
Novus Biologicals
NBP2-99091-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18636145
|
Novus Biologicals
NBP2-99091-50ul |
50 μg |
227.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
iNOS Polyclonal antibody specifically detects iNOS in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| iNOS | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Apoptosis, Cancer, HIF Target Genes, Hypoxia, Inflammation, Lipid and Metabolism, Neuroscience, Neurotransmission | |
| 0.2 μm filtered solution in PBS | |
| 4843 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/Immunofluorescence 1:300-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human iNOS. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title