missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Uniform Shape
- Low co-efficient of variation in terms of size
- High efficiency antibody binding for maximum sensitivity
- Available at high concentrations to simplify assay development
Specifications
Specifications
| Content And Storage | Store at 4°C. Do not freeze. |
| Quantity | 100 mL |
| Target | 40nm Colloidal Gold Nanoparticles 20 OD |
| Product Type | Antibody Labeling Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?