Learn More
Novus Biologicals™ 40nm Colloidal Gold Nanoparticles 20 OD
Product Code. 13511180 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13511180 100 mL Pack of 100
18036214 100 mL 100mL
18011386 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
For Research Use Only
Product Code. 13511180 Supplier Novus Biologicals™ Supplier No. 2200100
This item requires a price update prior to being ordered.
Request a Price for this item here.
For Research Use Only
High efficiency antibody binding for maximum sensitivity
  • Uniform Shape
  • Low co-efficient of variation in terms of size
  • High efficiency antibody binding for maximum sensitivity
  • Available at high concentrations to simplify assay development
Specifications
Content And Storage Store at 4°C. Do not freeze.
Quantity 100 mL
Target 40nm Colloidal Gold Nanoparticles 20 OD
Product Type Antibody Labeling Kit