missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INMT Antibody (S08-9C6), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00€ - 645.00€
Specifications
| Antigen | INMT |
|---|---|
| Clone | S08-9C6 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605903
|
Novus Biologicals
NBP3-19663-100ul |
100 μg |
645.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675023
|
Novus Biologicals
NBP3-19663-50ul |
50 μg |
421.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INMT Monoclonal antibody specifically detects INMT in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| INMT | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Amine N-methyltransferase, Aromatic alkylamine N-methyltransferase, Arylamine N-methyltransferase, EC 2.1.1, EC 2.1.1.49, Indolamine N-methyltransferase, indolethylamine N-methyltransferase, MGC125940, MGC125941, nicotine N-methyltransferase | |
| Recombinant protein of human INMT (Uniprot # O95050) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S08-9C6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 11185 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title