missing translation for 'onlineSavingsMsg'
Learn More

Inhibin alpha Mouse anti-Human, Clone: INHA/1168, Novus Biologicals™

Mouse Monoclonal Antibody

484.00€

Specifications

Antigen Inhibin alpha
Clone INHA/1168
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1 to 2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18665196
View Documents
Novus Biologicals
NBP3-07433-100ug
100 μg
484.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Inhibin alpha
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Apoptosis, Cancer
1 mM PBS with 0.05% BSA
3623
IgG2b κ
Protein A or G purified
INHA/1168
Immunohistochemistry-Paraffin 1 to 2 μg/mL
Monoclonal
Purified
RUO
Human
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha
Recombinant fragment of human Inhibin alpha protein (around aa 233-362) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.