missing translation for 'onlineSavingsMsg'
Learn More

Influenza A H1N1 PA Antibody - (A/Puerto Rico/8/1934), R&D Systems™

Product Code. 18148167 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18148167 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18148167 Supplier Novus Biologicals Supplier No. NBP242876
  • ONLINE EXCLUSIVE PRICE
    581.00€
    549.15€ / 0.1mL
    Save 31.85€
    5% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Influenza A H1N1 PA Polyclonal specifically detects Influenza A H1N1 PA in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H1N1 PA
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
Gene Alias Influenza PA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of Influenza A virus PA (A/Puerto Rico/8/1934(H1N1)). The exact sequence is proprietary.
Molecular Weight of Antigen 83 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.