missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Influenza A [A/Guinea fowl/Hong Kong/WF10/99 (H9N2)] Hemagglutinin 1 (HA1) Protein, His Tag
POA
Specifications
Accession Number | Q80KD9 |
---|---|
Molecular Weight (g/mol) | 37.5 kDa |
Source | Influenza A [A/Guinea fowl/Hong Kong/WF10/99 (H9N2)] HA1, His Tag (HA2-V52H8) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Arg 338 (Accession # Q80KD9). |
Immunogen | Asp 19 - Arg 338 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15816917
|
ACROBiosystems
HA2-V52H8-1MG |
1 mg |
POA
1mg |
N/A | |||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15806917
|
ACROBiosystems
HA2-V52H8-100UG |
100 µg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
Neuraminidase (NA) and hemagglutinin (HA) are major membrane glycoproteins found on the surface of influenza virus. Hemagglutinin binds to the sialic acid-containing receptors on the surface of host cells during initial infection and at the end of an infectious cycle. Hemagglutinin also plays a major role in the determination of host range restriction and virulence. As a class I viral fusion protein, hemagglutinin is responsible for penetration of the virus into the cell cytoplasm by mediating the fusion of the membrane of the endocytosed virus particle with the endosomal membrane.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.5 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
Q80KD9 | |
Influenza A [A/Guinea fowl/Hong Kong/WF10/99 (H9N2)] HA1, His Tag (HA2-V52H8) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Arg 338 (Accession # Q80KD9). | |
-20°C | |
Hemagglutinin 1 (HA1) | |
Influenza | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |
37.5 kDa | |
Asp 19 - Arg 338 | |
HA1,Hemagglutinin 1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |